Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HHV6 (U54) Peptide Pool

HHV6 (U54) Peptide Pool

1 AU
Katalog # STMC100-1401
Marketsource
Leverandør:   Stemcell Technologies
HHV6 (U54) Peptide Pool
Katalog # STMC100-1401
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
HHV6 (U54) Peptide Pool
HHV6 (U54) Peptide Pool

1 AU

CAS nummer:  
Price
4.564,00 kr
Log ind for tilgængelighed
1 * 1 YE
Product Details & Documents

Stimulating T cells with HHV6 (U54) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HHV6 (U54) peptide pool is a lyophilised mixture of 112 peptides from protein U54 of human herpesvirus 6B (HHV-6B; strain Z29), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 459 on U54. U54 is a tegument transactivator and is suggested to be a homolog of the CMV UL82/83 (pp65/72K) protein (Isegawa et al.; Nicholas). In infected T cells, U54 has been shown to inhibit gene transcription of interleukin 2 (IL-2), the T cell growth factor, which likely contributes to viral persistence (Iampietro et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1 AU
  • Cat. No.:
    STMC100-1401
  • Protein/peptide name:
    HHV6 (U54) Peptide Pool
  • Renhed:
    Average 70%

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.