Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
EBV (EBNA-1) Peptide Pool

EBV (EBNA-1) Peptide Pool

1 KIT
Katalog # STMC100-0669
Marketsource
Leverandør:   Stemcell Technologies
EBV (EBNA-1) Peptide Pool
Katalog # STMC100-0669
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
EBV (EBNA-1) Peptide Pool
EBV (EBNA-1) Peptide Pool

1 KIT

CAS nummer:  
Price
3.633,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

The EBV (EBNA-1) Peptide pool is a lyophilised mixture of 158 peptides from Epstein-Barr nuclear antigen 1 (EBNA-1) of Epstein-Barr virus (EBV; strain B95-8). EBNA-1 plays a critical role in the stable latent infection by EBV and may contribute to the survival of EBV-infected cells through its interaction with ubiquitin carboxyl-terminal hydrolase 7 (USP7). The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 641 on EBNA-1.

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1 KIT
  • Cat. No.:
    STMC100-0669
  • Protein/peptide name:
    EBV (EBNA-1) Peptide Pool
  • Renhed:
    Average 70%

Specifikationer