Order Entry
Denmark
ContactUsLinkComponent
Filter Paper, Glass Fibre, Extra Thick
Filter Paper, Glass Fibre, Extra Thick
Katalog # HACH2842100
Leverandør:  Hach
New Product
Filter Paper, Glass Fibre, Extra Thick
Katalog # HACH2842100
Leverandør:  Hach
New Product
Specifikationer
  • Beskrivelse:
    Glass microfibre filter paper, pore size: 1 µm, diameter: 47 mm
  • Størrelse:
    100 per pack
  • Pakn:
    100
Specifikationer
Om denne vare
For filtration of heavily contaminated samples. High particulate-holding capacity makes discs efficient depth filters and allows for filtration of large volumes of solutions.