Filter Paper, Glass Fibre, Extra Thick
Katalog # HACH2842100
Leverandør: Hach
New Product
Specifikationer
- Beskrivelse:Glass microfibre filter paper, pore size: 1 µm, diameter: 47 mm
- Størrelse:100 per pack
- Pakn:100
Specifikationer
Om denne vare
For filtration of heavily contaminated samples. High particulate-holding capacity makes discs efficient depth filters and allows for filtration of large volumes of solutions.