Orders LinkContactUsLinkComponent
UTTD Control Pharma Solution Homogenising System
UTTD control pharma solution
Katalog # IKAA10013297
Leverandør:   IKA
UTTD Control Pharma Solution Homogenising System
Katalog # IKAA10013297
Leverandør:  IKA
UTTD Control Pharma Solution Homogenising System
UTTD Control Pharma Solution Homogenising System
UTTD control pharma solution
Price
13.780,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Many standardised test procedures in the pharmaceutical industry, such as the pharmacopoeia tests described in official compendia (United States Pharmacopeia (USP 905), European Pharmacopoeia (PhEur 2.9.40), etc.), require the crushing and dissolution of tablets as a first step before further analysis. The challenges in sample preparation include the avoidance of cross-contamination between sample batches, time-consuming cleaning and the often small sample quantities.
  • Crushing and dissolving of tablets for subsequent analysis
The package includes the powerful ULTRA-TURRAX® Tube Drive P control homogenising system, which has been designed for universal use. In combination with the disposable mixing vessels with dissolver disc (DIS-300-S-M with 300 ml volume), this system ensures optimum comminution and thorough dissolution of tablets and capsules in various solvents.
The hermetically sealable disposable sample containers serve simultaneously as a comminution vessel, storage unit and transport container. They significantly reduce the cleaning effort and systematically eliminate cross-contamination. Users benefit from reduced handling costs and a lower number of incorrect analyses. Samples can be collected or media added via the pierceable membrane in the vessel lid.
 : ULTRA-TURRAX® Tube Drive P control, DIS-300-S-M.10 Dissolver tube, 300 ml, 10 pieces.
Specifications
  • Beskrivelse:
    UTTD control pharma solution
  • Cat. No.:
    IKAA10013297
  • Effekt:
    36 W
  • Hastighedsområde:
    400 - 8000 min⁻¹
  • Mål:
    122×178×54 mm
  • Maks. viskositet:
    5000 mPas
  • Vægt:
    1,3 kg
  • Hastighedsvisning:
    OLED
  • Motoreffekt input/output:
    36/28 W
Accessories
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • ST-50, rør med omrøring
    ST-50, rør med omrøring
    531-0480
    N
    50 ml
    Log in for availability
    1 * 10 Items - 910,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • BMT-20-S, rør til findeling, med rustfrit stål bolde
    BMT-20-S, rør til findeling, med rustfrit stål bolde
    531-0469
    N
    20 ml
    Log in for availability
    1 * 25 Items - 2.010,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • BMT-20 G kuglemøllerør med glaskugler
    BMT-20 G kuglemøllerør med glaskugler
    431-2893
    N
    20 ml
    Log in for availability
    1 * 25 Items - 2.010,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • DT-50, rør med rotor-stator element
    DT-50, rør med rotor-stator element
    531-0481
    N
    50 ml
    Log in for availability
    1 * 10 Items - 1.030,00 kr
Product Information
  • Katalog #
    Tilgængelighed
    Price
  • Glass beads for BMT tubes, Ø 6 mm
    Glass beads for BMT tubes, Ø 6 mm
    IKAA3599200
    Log in for availability
    1 * 250 g - 551,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • BMT-50-S-M-gamma, rør til findeling, med rustfrit stål bolde
    BMT-50-S-M-gamma, rør til findeling, med rustfrit stål bolde
    531-0493
    M
    50 ml
    Log in for availability
    1 * 10 Items - 1.750,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • BMT-50-G, rør til findeling med glas bolde
    BMT-50-G, rør til findeling med glas bolde
    531-0483
    N
    50 ml
    Log in for availability
    1 * 10 Items - 1.230,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • ST-20-M-gamma, rør med omrørings enhed
    ST-20-M-gamma, rør med omrørings enhed
    531-0477
    M
    20 ml
    Log in for availability
    1 * 20 Items - 2.040,00 kr
Product Information
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Labworldsoft® 6 Starter
    Labworldsoft® 6 Starter
    IKAA20019397
    Small configurations with up to 3 devices can be controlled at the same time
    Log in for availability
    1 * 1 Items - 45.630,00 kr
Product Information
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Labworldsoft® 6 Pro, with access to the entire device library
    Labworldsoft® 6 Pro, with access to the entire device library
    IKAA20017366
    All configurations with any number of devices can be controlled at the same time
    Log in for availability
    1 * 1 Items - 104.110,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • BMT-20-S-M-gamma, rør til findeling, med rustfrit stål bolde
    BMT-20-S-M-gamma, rør til findeling, med rustfrit stål bolde
    IKAA3700701
    M
    20 ml
    Log in for availability
    1 * 20 Items - 2.340,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • BMT-50-S, rør til findeling, med rustfrit stål bolde
    BMT-50-S, rør til findeling, med rustfrit stål bolde
    531-0482
    N
    50 ml
    Log in for availability
    1 * 10 Items - 1.230,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • DT-20 eco, rør med rotor/stator-element
    DT-20 eco, rør med rotor/stator-element
    IKAA20003207
    N
    Log in for availability
    1 * 25 Items - 1.630,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • ST-50-M-gamma, rør med omrørings enhed
    ST-50-M-gamma, rør med omrørings enhed
    531-0491
    M
    50 ml
    Log in for availability
    1 * 10 Items - 1.440,00 kr
Product Information
  • Katalog #
    Tilgængelighed
    Price
  • TC-20-M, Låg til 20 ml rør med preræble membran
    TC-20-M, Låg til 20 ml rør med preræble membran
    531-0496
    Log in for availability
    1 * 25 Items - 559,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • DT-50-M-gamma,rør med rotor-stator element
    DT-50-M-gamma,rør med rotor-stator element
    531-0492
    M
    50 ml
    Log in for availability
    1 * 10 Items - 1.630,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • DT-20-M-gamma, rør med rotor-stator element
    DT-20-M-gamma, rør med rotor-stator element
    531-0478
    M
    20 ml
    Log in for availability
    1 * 20 Items - 2.250,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • ST-20 omrøringsrør
    ST-20 omrøringsrør
    431-2892
    N
    20 ml
    Log in for availability
    1 * 25 Items - 1.820,00 kr
Product Information
  • Katalog #
    Tilgængelighed
    Price
  • Stainless steel (AISI 304) for BMT tubes, Ø 5 mm
    Stainless steel (AISI 304) for BMT tubes, Ø 5 mm
    IKAA3599300
    Log in for availability
    1 * 250 g - 551,00 kr
Product Information
  • Katalog #
    Tilgængelighed
    Price
  • TC-50-M, Låg til 50 ml rør med preræble membran
    TC-50-M, Låg til 50 ml rør med preræble membran
    431-0165
    Log in for availability
    1 * 10 Items - 368,00 kr
Product Information
  • Katalog #
    Lukkestørrelse
    Volumen
    Tilgængelighed
    Price
  • DT-20, rør med rotor/stator-element
    DT-20, rør med rotor/stator-element
    431-0135
    N
    20 ml
    Log in for availability
    1 * 25 Items - 1.900,00 kr