Orders LinkContactUsLinkComponent
Millenium Grip Antislip Overshoes
Millenium Grip Antislip Overshoes
Supplier:   MILLE S.A.S
Millenium Grip Antislip Overshoes
Supplier:  MILLE S.A.S
Certificates
Millenium Grip Antislip Overshoes
4 Options Available Below
Product Details & Documents
Millenium grip waterproof and anti-slip overshoes for wet and oily floor, ultra light and non-magnetic.
  • TPU waterproof overshoe
  • Easy on - easy off
  • Without toe-cap
  • Without penetration resistant midsole
  • Reusable, can by worn by more than 1 person => easy to wash
  • Exclusive design to avoid clogging with dirt
  • High performance grip, perfect for slippery, wet and oily floors
  • TPU outsole
  • Slip resistance: SRC
  • FO: Fuel oil resistant sole
  • Amagnetic (without metal)
  • Outsole abrasion and hydrocarbon resistant
  • Elasticity
  • Sole: Waterproof and washable TPU
Industry: Logistics, services, administrations, hygiene - food industry, catering.
Net weight per pair: 590 g.
Certifications: FO SRC, EN ISO 20344
Product Family Options
Product Information
  • Skostørrelse
    Farve
    Availability
    Price
  • 42/45
    sort
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 250,00 kr
    250,00 kr
    1 * 1 Items
       250,00 kr
  • 38/41
    sort
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 250,00 kr
    250,00 kr
    1 * 1 Items
       250,00 kr
  • 34/37
    sort
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 250,00 kr
    250,00 kr
    1 * 1 Items
       250,00 kr
  • 46/48
    sort
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 250,00 kr
    250,00 kr
    1 * 1 Items
       250,00 kr