Orders LinkContactUsLinkComponent
PEHANON® pH Indicator Test Strips
PEHANON® pH Indicator Test Strips
PEHANON® pH Indicator Test Strips
Leverandør:  MACHEREY-NAGEL
Certifikater
PEHANON® pH Indicator Test Strips
15 tilgængelige muligheder nedenfor
Product Details & Documents
PEHANON® are special pH test strips that unify the pH indicator and the reference colours on one strip. Any sample colour has the same effect on both the reference colours and the reactive pad. Users can be sure to get accurate readings.
  • Sample colour has no effect on the results
  • No accessories needed - always ready to use
  • Highly economic - colour scale on every single strip
  • Safe usage due to hydrophobic barrier
An invisible hydrophobic barrier above the top colour field prevents capillary action of the test solution, so the handle will remain dry and clean. pH values can be determined without a separate colour chart.
Certifications: Not CE certified.
Caution: No hazardous material.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 662-2329
  • 662-0218
  • 662-0214
  • MANA904930
  • 662-2330
  • 662-2331
  • 662-0215
  • 662-0117
  • 662-0216
  • 662-0217
  • 662-2332
  • 662-2333
  • 662-2334
  • 662-2335
  • 662-2336
  • Shelf life:
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
  • 3 year(s)
Specifikationer
Product Family Options
Produktinformation
  • Pakning
    Tilgængelighed
    Price
  • 662-2329
    PEHANON pH 0,0 - 1,8, 200 test strips
    PEHANON pH 0,0 - 1,8, 200 test strips
    Plastic box, 70×15×105 mm, 62,4 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2329
    Shelf life
    3 year(s)
  • 662-0218
    PEHANON pH 1,0 - 2,8, 200 test strips
    PEHANON pH 1,0 - 2,8, 200 test strips
    Plastic box, 70×15×106 mm, 62,08 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-0218
    Shelf life
    3 year(s)
  • 662-0214
    PEHANON pH 1,0 - 12,0, 200 test strips
    PEHANON pH 1,0 - 12,0, 200 test strips
    Plastic box, 70×15×100 mm, 62,28 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 205,00 kr
    205,00 kr
    1 * 200 Items
       205,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-0214
    Shelf life
    3 year(s)
  • MANA904930
    PEHANON pH 1,0 - 12,0
    PEHANON pH 1,0 - 12,0
    110×12×0 mm, 0,16 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 885,00 kr
    885,00 kr
    1 * 1.000 Items
       885,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA904930
    Shelf life
    3 year(s)
  • 662-2330
    PEHANON pH 1,8 - 3,8, 200 test strips
    PEHANON pH 1,8 - 3,8, 200 test strips
    Plastic box, 70×15×105 mm, 60,3 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2330
    Shelf life
    3 year(s)
  • 662-2331
    PEHANON pH 2,8 - 4,6, 200 test strips
    PEHANON pH 2,8 - 4,6, 200 test strips
    Plastic box, 70×15×105 mm, 61,51 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2331
    Shelf life
    3 year(s)
  • 662-0215
    PEHANON pH 3,8 - 5,5, 200 test strips
    PEHANON pH 3,8 - 5,5, 200 test strips
    Plastic box, 70×15×106 mm, 60 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-0215
    Shelf life
    3 year(s)
  • 662-0117
    PEHANON pH 4,0 - 9,0, 200 test strips
    PEHANON pH 4,0 - 9,0, 200 test strips
    Plastic box, 70×15×106 mm, 62,15 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 251,00 kr
    251,00 kr
    1 * 200 Items
       251,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-0117
    Shelf life
    3 year(s)
  • 662-0216
    PEHANON pH 5,2 - 6,8, 200 test strips
    PEHANON pH 5,2 - 6,8, 200 test strips
    Plastic box, 70×15×105 mm, 62 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-0216
    Shelf life
    3 year(s)
  • 662-0217
    PEHANON pH 6,0 - 8,1, 200 test strips
    PEHANON pH 6,0 - 8,1, 200 test strips
    Plastic box, 70×15×106 mm, 63,17 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-0217
    Shelf life
    3 year(s)
  • 662-2332
    PEHANON pH 7,2 - 8,8, 200 test strips
    PEHANON pH 7,2 - 8,8, 200 test strips
    Plastic box, 70×15×105 mm, 62,2 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2332
    Shelf life
    3 year(s)
  • 662-2333
    PEHANON pH 8,0 - 9,7, 200 test strips
    PEHANON pH 8,0 - 9,7, 200 test strips
    Plastic box, 70×15×100 mm, 61,14 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2333
    Shelf life
    3 year(s)
  • 662-2334
    PEHANON pH 9,5 - 12,0, 200 test strips
    PEHANON pH 9,5 - 12,0, 200 test strips
    Plastic box, 116×69×10 mm, 62 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2334
    Shelf life
    3 year(s)
  • 662-2335
    PEHANON pH 10,5 - 13,0, 200 test strips
    PEHANON pH 10,5 - 13,0, 200 test strips
    Plastic box, 70×15×105 mm, 61,22 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2335
    Shelf life
    3 year(s)
  • 662-2336
    PEHANON pH 12,0 - 14,0, 200 test strips
    PEHANON pH 12,0 - 14,0, 200 test strips
    Plastic box, 70×15×105 mm, 60,47 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 181,00 kr
    181,00 kr
    1 * 200 Items
       181,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    662-2336
    Shelf life
    3 year(s)