Orders LinkContactUsLinkComponent
Eosin B
Glass bottle for solids, 25 g
Katalog # SERA21003.01
Marketsource
Leverandør:   Serva
Eosin B
Katalog # SERA21003.01
Leverandør:  Serva
Marketsource
CAS nummer:  548-24-3
Eosin B
Glass bottle for solids, 25 g
CAS nummer:  548-24-3
Price
Spørg for pris
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
  • CAS nummer:548-24-3
  • Formel:C₂₀H₆Br₂N₂Na₂O₉
  • MDL nummer:MFCD00005041
  • Molecular Weight:624.06 g/mol
  • Merck Index:12,03638
  • Smeltende Pt:> 310 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-943-1
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g
Specifikationer