Neocuproine
Glass bottle for solids, 5 g
Catalog # SERA30230.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: 2,9-Dimethyl-1,10-phenanthroline
- CAS Number:484-11-7
- Formula:C₁₄H₁₂N₂
- MDL Number:MFCD00004973
- Molecular Weight:208.26 g/mol
- Merck Index:13,06477
- Melting Pt:161…163 °C
- Storage Temperature:Stue temperatur
- EINECS:207-601-9
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:5 g



