Orders LinkContactUsLinkComponent
BrainPhys™ Neuronal Medium
BrainPhys™ Neuronal Medium
BrainPhys™ Neuronal medium, 500 mL
Katalog # STMC05790
Marketsource
Leverandør:   Stemcell Technologies
BrainPhys™ Neuronal Medium
Katalog # STMC05790
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
BrainPhys™ Neuronal Medium
BrainPhys™ Neuronal Medium
BrainPhys™ Neuronal medium, 500 mL
CAS nummer:  
Price
913,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Serum-free neurophysiological basal medium for improved neuronal function.
Promote, rather than inhibit, neuronal activity and maturity in cultured primary or human pluripotent stem cell (hPSC)-derived neurons. Based on the formulation by Bardy and Gage (Bardy et al. PNAS, 2015), BrainPhys™ Neuronal Medium is a serum-free basal medium that is optimised to yield a higher proportion of synaptically active neurons by mimicking the central nervous system (CNS) extracellular environment. Use BrainPhys™ Neuronal Medium for long-term culture of hPSC- and CNS-derived neurons. To avoid shocking cells with media changes, also use BrainPhys™ medium when performing functional assays, such as microelectrode array-based recordings or live-fluorescent imaging.
To ensure cell health in long-term serum-free culture, BrainPhys™ Neuronal Medium must be combined with an appropriate serum-replacement supplement, such as NeuroCult™ SM1 Neuronal Supplement and/or N2 Supplement-A. For your convenience, various BrainPhys™ kits that include the required supplement(s) for primary or hPSC-derived neurons are also available. View our additional resources to learn more about the BrainPhys™ system.
Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    BrainPhys™ Neuronal medium