Orders LinkContactUsLinkComponent
MethoCult™ H4330 Medium
MethoCult™ H4330 Medium
MethoCult™ H4330, 90 mL
Katalog # STMC04330
Marketsource
Leverandør:   Stemcell Technologies
MethoCult™ H4330 Medium
Katalog # STMC04330
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
MethoCult™ H4330 Medium
MethoCult™ H4330 Medium
MethoCult™ H4330, 90 mL
CAS nummer:  
Price
4.884,00 kr
Log ind for tilgængelighed
1 * 90 mL
Product Details & Documents
Methylcellulose-based medium with EPO for human cells.
MethoCult™ H4330 is a methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. This formulation contains erythropoietin (EPO), but does not contain other cytokines. It is recommended for assays of late erythroid progenitors, specifically colony-forming cell - erythroid (CFU-E), in the presence of EPO only. It is also recommended for assays of early erythroid progenitors, specifically burst-forming unit - erythroid (BFU-E), and other colony-forming cells when appropriate cytokines are added. It can also be used to assess colony-stimulating factor activity or burst-promoting activity in unknown samples.
Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Pakn:
    90 mL
  • Beskrivelse:
    MethoCult™ H4330
Specifikationer