Orders LinkContactUsLinkComponent
ClonaCell™ HY Medium D
ClonaCell™ HY Medium D
ClonaCell™-HY Medium D, 90 mL
Katalog # STMC03804
Marketsource
Leverandør:   Stemcell Technologies
ClonaCell™ HY Medium D
Katalog # STMC03804
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
ClonaCell™ HY Medium D
ClonaCell™ HY Medium D
ClonaCell™-HY Medium D, 90 mL
CAS nummer:  
Price
5.405,00 kr
Log ind for tilgængelighed
1 * 90 mL
Product Details & Documents
Semi-solid methylcellulose-based medium for hybridoma selection and cloning, with HAT (serum-containing).
  • Forms physically separated, discrete colonies that can be easily isolated
  • Easily isolate rare and high-producing clones individually using simultaneous selection and cloning in semi-solid medium, compared with selection in bulk liquid cultures
  • Isolate monoclonal cell lines in less time and using fewer resources, compared with selection and cloning by limiting dilution
Save research time and resources by performing hybridoma selection and cloning in one step with this methylcellulose-based medium. Use ClonaCell™-HY Medium D to easily isolate individual clones with a high probability of monoclonality for further screening and expansion. Hybridoma clones typically have a wide range of growth rates and productivities; when cloned using ClonaCell™-HY Medium D, they form visible and discrete monoclonal colonies, preventing the loss of rare and high-producing clones due to overgrowth. The hybridoma colonies can be easily picked from the semi-solid medium by manual or robotic methods and dispersed into a liquid medium for screening and expansion.
ClonaCell™-HY Medium D is a semi-solid medium containing serum and the selective agents hypoxanthine, aminopterin, and thymidine (HAT). It has been verified for use in developing mouse and rat hybridomas. It has also been reported to be compatible with producing and cloning hybridomas using lymphocytes from a variety of host animals, including human, mouse, rat, and hamster (e.g. Wilson JR et al. Antiviral Res, 2016).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Pakn:
    90 mL
  • Beskrivelse:
    ClonaCell™-HY Medium D