Order Entry
Denmark
ContactUsLinkComponent
eTeSR™ Feeder-Free Medium
eTeSR™ Feeder-Free Medium
Catalog # STMC100-1215
CAS Number:  
undefined
eTeSR™ Feeder-Free Medium
Catalog # STMC100-1215
CAS Number:  
Specifications
  • Pakn:
    500 mL
  • Beskrivelse:
    eTeSR™ feeder-free medium
Specifications
About this item
Stabilised, feeder-free maintenance medium for human ES and iPS cells optimised for single-cell passaging.
Improve the genetic stability of human pluripotent stem cell (hPSC) cultures when performing routine single-cell passaging by using eTeSR™, an enhanced feeder-free culture medium. eTeSR™ builds upon previous formulations of TeSR™, the most widely published feeder-free cell culture media for hPSCs. Specifically developed for single-cell applications, which typically involve shorter passaging schedules at high density, eTeSR™ can be used for routine maintenance, gene editing, or cloning workflows. Use eTeSR™ to produce high-quality hPSCs with improved genetic stability for these applications; stabilisation of the components (e.g. FGF2), optimised metabolites, and improved buffering capacity address the increased metabolic demand and cell stress associated with single-cell passaging. Expect equivalent high performance when using eTeSR™ with either daily or restricted feeding schedules. eTeSR™ is compatible with a variety of cell culture matrices, including Corning® Matrigel® hESC-Qualified Matrix and CellAdhere™ Laminin-521. Each lot of eTeSR™ 10X Supplement is quality-tested in a culture assay using hPSCs.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').