Order Entry
Denmark
Orders LinkContactUsLinkComponent
MethoCult™-GF M3434 Medium

MethoCult™-GF M3434 Medium

MethoCult™ GF M3434, 100 mL
Katalog # STMC03434
Marketsource
Leverandør:   Stemcell Technologies
MethoCult™-GF M3434 Medium
Katalog # STMC03434
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
MethoCult™-GF M3434 Medium
MethoCult™-GF M3434 Medium

MethoCult™ GF M3434, 100 mL

CAS nummer:  
Price
6.037,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Methylcellulose-based medium with recombinant cytokines (including EPO) for mouse cells. 1×100 ml bottle.

MethoCult™ GF M3434 is optimised for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver cells. MethoCult™ GF M3434 has been formulated to support optimal growth of primitive erythroid progenitor cells (BFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multi-potential granulocyte, erythroid, macrophage, megakaryocyte progenitor cells (CFU-GEMM). This formulation is compatible with STEMvision™ software for automated colony counting of mouse bone marrow CFU assays.

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    100 mL
  • Beskrivelse:
    MethoCult™ GF M3434

Specifikationer