Orders LinkContactUsLinkComponent
L-Glutamine
L-Glutamine
L-Glutamine, 100 mL
Katalog # STMC07100
Marketsource
Leverandør:   Stemcell Technologies
L-Glutamine
Katalog # STMC07100
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
L-Glutamine
L-Glutamine
L-Glutamine, 100 mL
CAS nummer:  
Price
602,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Cell culture supplement. 1×3 ml vial.
L-Glutamine (200 mM) is recommended for use as a cell culture supplement and is required for the preparation of MesenCult™-ACF plus medium.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Pakn:
    100 mL
  • Beskrivelse:
    L-Glutamine