Anti-LRRN3 Rabbit Polyclonal Antibody
400 µl
Catalog # PRSI56-149
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
LRRN3 is a single pass type I membrane protein. It contains 1 fibronectin type III domain, 1 Ig like C2 type (immunoglobulin like) domain and 12 LRR (leucine rich) repeats. The function of the LRRN3 protein remains unknown.
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: LRRN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Specifications
- Størrelse:400 µl
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Gen ID:Q9H3W5
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Konjugation:Unconjugated
- Form:Liquid
- Antigen synonymer:LRRN3
- Koncentration:0,5 mg/ml
- Epitop:Amino Acid Range = 596-623
- Shipping temperature:4 °C
- Antigen name:Leucine-rich repeat neuronal protein 3
- Cat. No.:PRSI56-149
- Reactivity:Human
- Molekylvægt:79 kDa
- Immunogen:This LRRN3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 596-623 amino acids from the C-terminal region of human LRRN3.
- Vært:Rabbit
- Antistof type:Primary
- Opbevaringstemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Antigen symbol:LRRN3
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



