Anti-NXT2 Rabbit Polyclonal Antibody
Katalog # PRSI15-219
Supplier: ProSci Inc.
Specifications
- Antistof type:Primary
- Antigen name:Nuclear transport factor 2 like export factor 2
- Antigen symbol:NXT2
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gen ID:55916
- Antigen synonymer:P15-2|NTF2-related export protein 2|protein p15-2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Opbevaringstemperatur:Store at −20 °C. Avoid freeze/thaw cycles
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 110-180 of human NXT2 (NP_061168.2)
- Purification:Affinity purification
- Pakning:Vial
- Pakn:50 µl
Specifications
About this item
Type: Primary
Antigen: NXT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat