Anti-PPP3CA Rabbit Polyclonal Antibody
Katalog # PRSI63-485
Leverandør: ProSci Inc.
Gå til:
Product Details & Documents
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: PPP3CA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 21-51
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse
Specifikationer
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Gen ID:5530
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Konjugation:Unconjugated
- Form:Liquid
- Koncentration:2 mg/ml
- Epitop:21-51 aa
- Shipping temperature:4 °C
- Antigen name:Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
- Cat. No.:PRSI63-485
- Reactivity:Human|Mouse
- Molekylvægt:59 kDa
- Immunogen:This Calcineurin (PPP3CA) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 21-51 amino acids from the N-terminal region of human Calcineurin (PPP3CA).
- Vært:Rabbit
- ImmunoChemistry:Yes
- Antistof type:Primary
- Opbevaringstemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Antigen symbol:PPP3CA



