Cooling Evaporative Vests
Om denne vare
Manufactured with super absorbent fibres, this vest is designed to keep workers cool for hours of extended wear.
- Lightweight and comfortable
- Cooling mesh fabric for increased breathability
- Keeps wearer cool for up to 8 hours
- Wicking, cooling, and drying capabilities
- Soak in water to activate
Lightweight and easy to activate, the Cooling Evaporative Vest will keep the wearer cool without wetting the clothes worn underneath.
Recommended for use in environments requiring high visibility and reflective clothing. Yellow finish.
: EN 17353 Type A. CE certified.
3 tilgængelige muligheder nedenfor
Product Details & Documents
Manufactured with super absorbent fibres, this vest is designed to keep workers cool for hours of extended wear.
- Lightweight and comfortable
- Cooling mesh fabric for increased breathability
- Keeps wearer cool for up to 8 hours
- Wicking, cooling, and drying capabilities
- Soak in water to activate
Lightweight and easy to activate, the Cooling Evaporative Vest will keep the wearer cool without wetting the clothes worn underneath.
Recommended for use in environments requiring high visibility and reflective clothing. Yellow finish.
: EN 17353 Type A. CE certified.
Specifications for Product Family
Specifikationer
- Catalog No:
- PRTWCV09YERS/M
- PRTWCV09YERL/XL
- PRTWCV09YERXX/3X
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- HiVis
- HiVis
- HiVis
- Køn:
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 100% polyester
- 100% polyester
- 100% polyester
- Vægt:
- 0,321 g/m²
- 0,366 g/m²
- 0,391 g/m²
Specifikationer
Product Family Options
Produktinformation
- FarveTøj størrelseBeklædningsgenstandTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.PRTWCV09YERS/MOutfit applikationerOutdoorRisk areaHiVisKønUnisexSleeve or leg lengthShortLukkestørrelseMed lynlåsStof100% polyesterVægt0,321 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWCV09YERL/XLOutfit applikationerOutdoorRisk areaHiVisKønUnisexSleeve or leg lengthShortLukkestørrelseMed lynlåsStof100% polyesterVægt0,366 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWCV09YERXX/3XOutfit applikationerOutdoorRisk areaHiVisKønUnisexSleeve or leg lengthShortLukkestørrelseMed lynlåsStof100% polyesterVægt0,391 g/m²