Cedertræsolie
Glass bottle, 100 mL
Katalog # ROTH5308.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Cedar wood oil, Cedar Oil, Cedar Leaf Oil
- CAS nummer:8000-27-9
- MDL nummer:MFCD00146327
- Tæthed:0,98 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



