Orders LinkContactUsLinkComponent
Congo red, Supelco®
Glass bottle for solids, 25 mg
Katalog # 75768-25MG
Sundhed
Leverandør:   Merck
Congo red, Supelco®
Katalog # 75768-25MG
Leverandør:  Merck
CAS nummer:  573-58-0
Congo red, Supelco®
Congo red, Supelco®
Glass bottle for solids, 25 mg
CAS nummer:  573-58-0
Price
1.100,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents
Synonymer:  CI 22120, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalen-1-sulfonat)
  • CAS nummer:573-58-0
  • Formel:C₃₂H₂₂N₆Na₂O₆S₂
  • MDL nummer:MFCD00004028
  • Molecular Weight:696.67 g/mol
  • Merck Index:12,02562
  • Smeltende Pt:> 360 °C
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-358-4
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 mg