Plastic Box with White Lid
Plastic box with safety lock and white lid, 3 L, 193×193×112 mm, PP
Katalog # MULC25685
Leverandør: MULTIROIR
Gå til:
Product Details & Documents
Box opaque with airtight and tamper-evident lid, perfect for use in laboratories.
- Polypropylene material
- Opaque white
- Watertight lid
- Tamper-proof lid
- Stackable box
- Food contact quality
Ideal for laboratory environments, this versatile container ensures the secure storage and transport of samples and reagents. The airtight, tamper-evident lid provides enhanced safety, while the stackable design maximises space efficiency. The opaque white polypropylene construction ensures durability and easy identification of contents.
Documents
MULC25685_TDS_EN
Datablad
Specifikationer
- Kapacitet:3 L
- Beskrivelse:Plastic box with safety lock and white lid
- Materiale:PP
- B×D×H:193×193×112 mm