Gas Washing Bottle, Desaga Pattern
Wash bottle round bottom, single, metrics: 34×150 mm, 100 ml, GL 25
Katalog # NEUB0578-10-010025
Leverandør: NEUBERT VOLUME GLASSWAERE
Gas Washing Bottle, Desaga Pattern
Katalog # NEUB0578-10-010025
Leverandør: NEUBERT VOLUME GLASSWAERE
Gå til:
Product Details & Documents
Bautele individually, from Borosilicate galss 3.3, inserts with safety glass ring.
Specifikationer
- Gevind:GL 25
- Kapacitet:100 ml
- Beskrivelse:Wash bottle round bottom, single, metrics: 34×150 mm