Orders LinkContactUsLinkComponent
Jod 0,5M (1N)
Jod 0,5M (1N)
1 L
Catalog # ACSDZ13501035.1L
Irritant
Supplier:   CPACHEM
Jod 0,5M (1N)
Catalog # ACSDZ13501035.1L
Supplier:  CPACHEM
CAS Number:  7553-56-2
Jod 0,5M (1N)
1 L
CAS Number:  7553-56-2
Price
3.300,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:7553-56-2
  • Formula:I₂
  • MDL Number:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Boiling Pt:100 °C
  • Density:~1,92511 g/cm³
Specifications
Specifications
  • Pakn:
    1 L