


4-Nonylphenol, ethoxylated
Catalog # ACSDSB37640.100MG
Supplier: CPACHEM
CAS Number:
Specifications
- Pakn:100 mg
- Seq:2440
Specifications
About this item
:
Melting Pt:57 °C
Density:1,6 g/cm³ at 20 °C
Flash Pt:>93 °C
UN:3082
ADR:6.1,III
:GHS05
:GHS07
:GHS09
CAS Number:9016-45-9
MDL Number:MFCD00132851