Orders LinkContactUsLinkComponent
WHEATON® FTA® Nucleic Acid Collection Storage Boxes
WHEATON® FTA® Nucleic Acid Collection Storage Boxes
Leverandør:   DWK Life Sciences
WHEATON® FTA® Nucleic Acid Collection Storage Boxes
Leverandør:  DWK Life Sciences
Certifikater
WHEATON® FTA® Nucleic Acid Collection Storage Boxes
2 tilgængelige muligheder nedenfor
Product Details & Documents
Storage boxes provide an ideal method for batching/storing FTA® Nucleic acid collection pouches.
  • Water repellent fiberboard box material allows for longer durability
  • For use with small or large FTA cards
  • Accommodates two rows of 76,2 mm FTA card pouches or one row of 152,4 FTA card pouches
  • Removable divider for use with smaller pouches
  • For use at RT, +4, −20 °C or colder
Storage boxes are ideal for nucleic acid collection.
Product Family Options
Produktinformation
  • B×D×H
    Tilgængelighed
    Price
  • 457,2×152,4×101,6 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 390,00 kr
    390,00 kr
    1 * 2 Items
       390,00 kr
  • 457,2×177,8×101,6 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 Items
    1 * 4 Items - 758,00 kr
    758,00 kr
    1 * 4 Items
       758,00 kr