Butyrometer original Gerber
Om denne vare
DURAN®, borosilicate glass.
Butyrometers are used to determine the fat content of dairy samples. The product is treated with concentrated sulphuric acid to disrupt the emulsion. The resulting mixture is centrifuged in the butyrometer and the fat content read directly off the scale.
5 tilgængelige muligheder nedenfor
Product Details & Documents
DURAN®, borosilicate glass.
Butyrometers are used to determine the fat content of dairy samples. The product is treated with concentrated sulphuric acid to disrupt the emulsion. The resulting mixture is centrifuged in the butyrometer and the fat content read directly off the scale.
Product Family Options
Accessories
Produktinformation
Produktinformation
- TilgængelighedPrice
- 709-0132Butyrometer til mælk, 0 - 4 %:0,05, ISO 396c - 4
- 709-0133Butyrometer til mælk, 0 - 8 %:0,1, ISO 510 - 8
- 709-0134Butyrometer til ost, 0-40 %:0,5, ISO/DIN/NE 239 - 40Butyrometer til ost, 0-40 %:0,5, ISO/DIN/NE 239 - 40Log ind for tilgængelighed1 * 1 Items694,00 kr
- 709-0135Butyrometer til ost, 0 - 40 %:0,5, ISO/DIN/NE 239 - 40, uden glasbægerButyrometer til ost, 0 - 40 %:0,5, ISO/DIN/NE 239 - 40, uden glasbægerLog ind for tilgængelighed1 * 1 Items497,00 kr