Orders LinkContactUsLinkComponent
VHB DNA Washing Buffer
VHB DNA Washing Buffer
Supplier:   OMEGA BIO-TEK
VHB DNA Washing Buffer
Supplier:  OMEGA BIO-TEK
Certificates
VHB DNA Washing Buffer
2 Options Available Below
Product Details & Documents
VHB buffer must be diluted with ethanol before use.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • Beskrivelse
    Pakn
    Availability
    Price
  • VHB buffer
    440 mL
    Log in for availability
    Log in for availability
    1 * 440 mL
    1 * 440 mL - 4.690,00 kr
    4.690,00 kr
    1 * 440 mL
       4.690,00 kr
  • VHB buffer
    1.760 mL
    Log in for availability
    Log in for availability
    1 * 1.760 mL
    1 * 1.760 mL - 17.860,00 kr
    17.860,00 kr
    1 * 1.760 mL
       17.860,00 kr