Orders LinkContactUsLinkComponent
WHEATON® PIPET-PAL® Pipette Controller, Translucent Blue
WHEATON® PIPET-PAL® Pipette Controller, Translucent Blue
PIPET-PAL™ pipette controller, gennemsigtig blå
Katalog # 612-6770
Leverandør:   DWK Life Sciences
WHEATON® PIPET-PAL® Pipette Controller, Translucent Blue
Katalog # 612-6770
Leverandør:  DWK Life Sciences
WHEATON® PIPET-PAL® Pipette Controller, Translucent Blue
PIPET-PAL™ pipette controller, gennemsigtig blå
Price
6.730,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PIPET-PAL® pipette controller are lightweight (195 g) and well-balanced, making even prolonged pipetting very comfortable.
  • Quiet, robust pump for high pipetting speed
  • Versatile for both fine dosing and gravimetric dispensing
  • Eliminate contamination with a replaceable, sterile filter
  • Lithium-Polymer battery technology for long operating time
  • Autoclavable pipette holder for cleanliness and safety
  • One year warranty
Precise finger-control of pipetting enables accurate dispensing, even drop-by drop.
Designed for use with all serological pipettes in the volume range 1 to 100 ml.
Specifikationer
  • Farvekode:
    gennemsigtig blå
  • Beskrivelse:
    PIPET-PAL™ pipette controller
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement filters 0,20 µm, PTFE, sterile
    Replacement filters 0,20 µm, PTFE, sterile
    612-6775
    Log ind for tilgængelighed
    1 * 1 Items - 1.630,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement filters 0,20 µm, PTFE, non sterile
    Replacement filters 0,20 µm, PTFE, non sterile
    612-6774
    Log ind for tilgængelighed
    1 * 10 Items - 1.380,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement filters 0,45 µm, PTFE, non sterile
    Replacement filters 0,45 µm, PTFE, non sterile
    612-6772
    Log ind for tilgængelighed
    1 * 10 Items - 1.090,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement filters 0,45 µm, PTFE, sterile
    Replacement filters 0,45 µm, PTFE, sterile
    612-6773
    Log ind for tilgængelighed
    1 * 10 Items - 1.360,00 kr