Orders LinkContactUsLinkComponent
Crockery Basket
Crockery Basket
Crockery basket, 500×500×133 mm
Katalog # HUPF0115332
Leverandør:   HUPFER METALLWERKE
Crockery Basket
Katalog # HUPF0115332
Leverandør:  HUPFER METALLWERKE
Crockery Basket
Crockery basket, 500×500×133 mm
Price
1.310,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Crockery basket for storage and transport of small or difficult-to-stack crockery items in basket dispenser or basket transport trolley.
  • Hygienic design
  • Made of high-quality, electro-polished stainless steel round material in loop design
  • Weight: 1,647 kg
Features: Basket frame 5 mm, basket base and walls 2,5 mm, the mesh size is 27 mm within each loop and 30 mm from loop to loop, 133 mm high, Ø 4 mm centring corners integrated into the frame, for impeccable and precise stacking.
Specifikationer
  • Beskrivelse:
    Crockery basket
  • B×D×H:
    500×500×133 mm