Chairs, Neon 3
Lab chair with foot rest, 590 - 870 mm, Stop and go castors, With foot rest
Katalog # BIMO95712000327780
Leverandør: bimos
Gå til:
Product Details & Documents
Neon 3 chairs comes with mounting aid and glides.
- Ergonomic package
- Seat height adjustment
- Seat inclination adjustment
- Seat depth adjustment
- Backrest height adjustment
- Aluminium base (black optional)
Mounting aid is height adjustable and folded up.
Backrest shell: Basalt grey
Specifikationer
- Beskrivelse:Lab chair with foot rest
- Højde:590 - 870 mm
- Størrelse:Stop and go castors
- Fodstøttering:With foot rest
- Cat. No.:BIMO95712000327780
- Seat/back color:Ocean blue/black
- Model:Neon 3