Orders LinkContactUsLinkComponent
Vacu-vials® Snapper
Vacu-vials® Snapper
Vacu-vials snapper
Katalog # CHETA-0214
Leverandør:   CHEMetrics
Vacu-vials® Snapper
Katalog # CHETA-0214
Leverandør:  CHEMetrics
Vacu-vials® Snapper
Vacu-vials snapper
Price
827,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This reusable vial snapper permits beverage manufacturers to conveniently monitor disinfectant and sterilisation residual levels directly inside a bottle or carton taken off the production line. Since the measurement is obtained in a single container, sample agitation is minimised leading to more accurate measurements.
Specifikationer
  • Beskrivelse:
    Vacu-vials snapper