Soxhlet Extraction Apparatus
Soxhlet extraction apparatus with standard taper ground joints, 250 ml
Katalog # WHEA416005
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Soxhlet extraction apparatus with standard taper ground joints is used for continuous extraction of analytes from a solid into an organic solvent.
- Manufactured from low extractable borosilicate glass
As the flask containing the solvent is heated, vapors rise in the larger outside tube, enter the water-cooled condenser and liquate and when the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask.
Certifications: Conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Specifikationer
- Kapacitet:250 ml
- Beskrivelse:Soxhlet extraction apparatus with standard taper ground joints
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Flat-bottom flask, 125 ml, 24/40