ContactUsLinkComponent
Ammoniumthiocyanat 0.1 M NH4SCN (0.1N) volumetrisk opløsning, Fluka™
Ammoniumthiocyanat 0.1 M NH4SCN (0.1N) volumetrisk opløsning, Fluka™
Catalog # 35036-1L
CAS Number:  
Ammoniumthiocyanat 0.1 M NH4SCN (0.1N) volumetrisk opløsning, Fluka™
Catalog # 35036-1L
CAS Number:  
Specifications
  • Pakning:
    Glass bottle
  • Pakn:
    1 L
  • Seq:
    5037
Specifications
Specification Test Results
  • Factor:
    0.998 - 1.002
Specification Test Results
About this item
 :
 :76.12 g/mol
Boiling Pt:100 °C (1,013 hPa)
Storage Temperature:Stue temperatur
EINECS:231-791-2
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428