Steep Breast Bottle
1000 ml (1 L), Steep breast bottle, 29/32, NS-8-edge stopper, EH
Katalog # WITG5081001
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Steep breast bottles, soda glass with safety coating WITOSINT.
Specifikationer
- NS:29/32, NS-8-edge stopper, EH
- Kapacitet:1000 ml (1 L)
- Beskrivelse:Steep breast bottle