Orders LinkContactUsLinkComponent
L-Alanin
Marketsource
 :   Serva
L-Alanin
 :  Serva
Marketsource
L-Alanin
 :  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  •  :56-41-7
  •  :C₃H₇NO₂
  •  :MFCD00064410
  •  :89.09 g/mol
  •  :13,00203
  •  :189 °C (1013 hPa)
  •  :79 °C
  •  :1,4 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :200-273-8
Product Family Options
  • Pakning
    Pakn
  • Glass bottle for solids
    25 g
    1 * 25 g
    1 * 25 g
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    1 * 100 g
    1 * 100 g
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g