Orders LinkContactUsLinkComponent
ActivArmr® Electrical Insulating Gloves, Class 4
ActivArmr® Electrical Insulating Gloves, Class 4
Leverandør:   Ansell
ActivArmr® Electrical Insulating Gloves, Class 4
Leverandør:  Ansell
Certifikater
ActivArmr® Electrical Insulating Gloves, Class 4
ActivArmr® Electrical Insulating Gloves, Class 4
8 tilgængelige muligheder nedenfor
Product Details & Documents
Durable, form-fitting electrical safety gloves, offering flexible, comfortable hand protection. Arc-flash resistant.
  • Natural rubber latex for ultimate durability and flexibility
  • Ergonomically-designed hand-at-rest shape for less hand fatigue
  • Generous flared cuff allows room for clothing and improves ventilation
  • Case hardened finish provides smoother surface for easy donning and doffing performance and protection
  • Product meets all applicable NFPA, OSHA and CSA standards
  • Washing temperature: 60 °C
  • Liner colour: Yellow
Recommended for: Applications with risks that involve heat, cuts and abrasions, electrical contractors, hybrid engine manufacturing and repair, mass transit repair and maintenance, power maintenance, repair and transmission, telecom repair near electrical wires.
Ideal for use in the following industries: automotive, construction, electrical, infrastructure maintenance.
Certifications: EN 388:2016 (X243D), EN 60903:2003, ARC Rating of APC 2, ASTM D120, ASTM F2675:2021, Open Arc: EBT: 88,2 cal/cm²; Cat. III
Packaging: Gloves are packaged 1 pair per bag, 10 pairs per carton.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • ANSERIG418YBCC090
  • ANSERIG418YBCC100
  • ANSERIG418YBCC110
  • ANSERIG418YBCC120
  • ANSERIG418YBSC090
  • ANSERIG418YBSC100
  • ANSERIG418YBSC110
  • ANSERIG418YBSC120
  • Model:
  • RIG418YBCC
  • RIG418YBCC
  • RIG418YBCC
  • RIG418YBCC
  • RIG418YBSC
  • RIG418YBSC
  • RIG418YBSC
  • RIG418YBSC
  • Ambidextrous/Hånd-specifik:
  • Hånd-specifik
  • Hånd-specifik
  • Hånd-specifik
  • Hånd-specifik
  • Hånd-specifik
  • Hånd-specifik
  • Hånd-specifik
  • Hånd-specifik
  • Engangs/Genbrugelig:
  • Genanvendelig
  • Genanvendelig
  • Genanvendelig
  • Genanvendelig
  • Genanvendelig
  • Genanvendelig
  • Genanvendelig
  • Genanvendelig
  • Latex/latex-fri:
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Materiale:
  • Naturgummilatex
  • Naturgummilatex
  • Naturgummilatex
  • Naturgummilatex
  • Naturgummilatex
  • Naturgummilatex
  • Naturgummilatex
  • Naturgummilatex
  • Overflade:
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Manchet:
  • Rullet kant
  • Rullet kant
  • Rullet kant
  • Rullet kant
  • Rullet kant
  • Rullet kant
  • Rullet kant
  • Rullet kant
  • Cuff længde:
  • 460 mm
  • 460 mm
  • 460 mm
  • 460 mm
  • 460 mm
  • 460 mm
  • 460 mm
  • 460 mm
  • EN norm:
  • EN 388:2016 (X243D), EN 60903:2003
  • EN 388:2016 (X243D), EN 60903:2003
  • EN 388:2016 (X243D), EN 60903:2003
  • EN 388:2016 (X243D), EN 60903:2003
  • EN 388:2016 (X243D), EN 60903:2003
  • EN 388:2016 (X243D), EN 60903:2003
  • EN 388:2016 (X243D), EN 60903:2003
  • EN 388:2016 (X243D), EN 60903:2003
Specifikationer
Product Family Options
Contour cuff with rolled beaded edge
Produktinformation
  • Model
    Handske størrelse
    Farve
    Længde
    Tilgængelighed
    Price
  • RIG418YBCC
    9
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBCC090
    Model
    RIG418YBCC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003
  • RIG418YBCC
    10
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBCC100
    Model
    RIG418YBCC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003
  • RIG418YBCC
    11
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBCC110
    Model
    RIG418YBCC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003
  • RIG418YBCC
    12
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBCC120
    Model
    RIG418YBCC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003
Straight cuff with rolled beaded edge
Produktinformation
  • Model
    Handske størrelse
    Farve
    Længde
    Tilgængelighed
    Price
  • RIG418YBSC
    9
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par - 434,00 kr
    434,00 kr
    1 * 1 Par
       434,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBSC090
    Model
    RIG418YBSC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003
  • RIG418YBSC
    10
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par - 434,00 kr
    434,00 kr
    1 * 1 Par
       434,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBSC100
    Model
    RIG418YBSC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003
  • RIG418YBSC
    11
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par - 434,00 kr
    434,00 kr
    1 * 1 Par
       434,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBSC110
    Model
    RIG418YBSC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003
  • RIG418YBSC
    12
    sort
    460 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Par
    1 * 1 Par - 434,00 kr
    434,00 kr
    1 * 1 Par
       434,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    ANSERIG418YBSC120
    Model
    RIG418YBSC
    Ambidextrous/Hånd-specifik
    Hånd-specifik
    Engangs/Genbrugelig
    Genanvendelig
    Latex/latex-fri
    Latex
    Materiale
    Naturgummilatex
    Overflade
    Smooth
    Manchet
    Rullet kant
    Cuff længde
    460 mm
    EN norm
    EN 388:2016 (X243D), EN 60903:2003