
Sodium pyruvate-¹³C₃ 99%
Catalog # CAMBCLM-2240-1
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pakn:1 g
- Seq:3047
Specifications
Specification Test Results
- Concentration:Neat
- pH:7
- Solubility:Water: 100 %
Specification Test Results
About this item
:
:113.02 g/mol
Melting Pt:>300 °C
:GHS07
Storage Temperature:Køleskab
CAS Number:142014-11-7
Formula:¹³CH₃¹³CO¹³CO₂Na
MDL Number:MFCD00084084