Natriummetabisulfit
10 g
Catalog # PROOTRC-S66706010G
Supplier: LGC Standards PROMOCHEM
Natriummetabisulfit
Catalog # PROOTRC-S66706010G
Supplier: LGC Standards PROMOCHEM
CAS Number: 7681-57-4
Jump to:
Product Details & Documents
Caution: Use of water spray when fighting fire may be inefficient.
Synonyms: Natriumdisulfit, di-Natriumdisulfit, Natriumpentaoxodisulfat
- CAS Number:7681-57-4
- Formula:Na₂S₂O₅
- MDL Number:MFCD00167602
- Molecular Weight:190.11 g/mol
Specifications
Specification Test Results
- Appearance:White Solid
- MS:Conforms
- Sodium Content:24.6%
Specifications
- Pakn:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



