Orders LinkContactUsLinkComponent
Multi-Parameter Tester, CombiCheck 10
Multi-Parameter Tester, CombiCheck 10
CombiCheck 10 Muliti-parameter standard solutions, 1 KIT
Katalog # 713-4096
Leverandør:   WTW
Multi-Parameter Tester, CombiCheck 10
Katalog # 713-4096
Leverandør:  WTW
CAS nummer:  
Multi-Parameter Tester, CombiCheck 10
Multi-Parameter Tester, CombiCheck 10
CombiCheck 10 Muliti-parameter standard solutions, 1 KIT
CAS nummer:  
Price
1.200,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
CombiCheck solutions are ready-to-use multi-parameter standards, Each package contains a standard solution and an addition solution. Both solutions can be used directly without dilution for quality assurance.
  • The standard solution is used to check the accuracy of the results for the complete system: procedure - analytical method - reagents - photometer
  • The addition solution is used to check sample-dependent influences by measuring the recovery rate and to determine the most suitable sample preparation method
The maximum number of determinations which can be made with a CombiCheck standard solution depends on the test set used. With the addition solution 280 determinations are always possible.
Parameters can be checked with their concentrations as follows:
Ammonium: 4,00 mg/L NH₄-N
Chloride: 25,0 mg/L Cl
COD: 80 mg/L COD
Nitrate: 2,5 mg/L NO₃-N
Phosphate: 0,80 mg/L PO₄-P
Sulphate: 100 mg/L SO₄
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    CombiCheck 10 Muliti-parameter standard solutions