Orders LinkContactUsLinkComponent
illustra™ TempliPhi™ sequence resolver kit
illustra™ TempliPhi™ sequence resolver kit
Leverandør:   Cytiva
illustra™ TempliPhi™ sequence resolver kit
Leverandør:  Cytiva
Certifikater
illustra™ TempliPhi™ sequence resolver kit
illustra™ TempliPhi™ sequence resolver kit
2 tilgængelige muligheder nedenfor
Product Details & Documents
For the amplification of difficult templates for successful DNA sequencing in 18 hours.
  • Easy-to-use kit requires only 20 minutes hands-on time: Solve the most common sequencing problems such as repeats, sequencing stops, and compressions with a single kit
  • Cultureless preparation from bacterial colonies, glycerol stock or purified DNA; saves time and reagents
  • Improve difficult template sequencing success with up to 820 bp Phred 20 read length
illustra TempliPhi Sequence Resolver Kit produces exceptional sequencing results from difficult templates, such as those with high GC content and secondary structures and uses the highly processive, strand-displacing Phi29 DNA polymerase and modified nucleotides to amplify circular templates for subsequent sequencing.
The kit resolves most of the sequencing stops or gaps that elude other used finishing methods. It can be used with both small (plasmids and M13) and large (fosmids and bacterial artificial chromosomes [BACs]) circular templates. The starting template for the reaction can be purified DNA, glycerol stock, liquid culture, or colonies. After an overnight incubation at 10 °C, it is ready to be used in sequencing applications with the sequencing chemistry of choice. A 10 μl reaction yields 1 to 2 μg of purified template DNA in a standard overnight reaction.
Bestillingsoplysninger: Sample buffer (white cap), reaction buffer (blue cap), and enzyme mix (yellow cap). Includes a certificate of analysis.
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 28-9035-29
  • 28-9035-31
  • Opbevaring betingelser:
  • Enzyme mix to be stored at -70°C; all other components at -20°C.
    Thaw components on ice at 0 to 4°C during handling
  • Enzyme mix to be stored at -70°C; all other components at -20°C.
    Thaw components on ice at 0 to 4°C during handling
Specifikationer
Product Family Options
Produktinformation
  • No. of reactions
    Pakn
    Tilgængelighed
    Price
  • 28-9035-29
    illustra™ TempliPhi™ Sequence Resolver Kit
    illustra™ TempliPhi™ Sequence Resolver Kit
    20
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.600,00 kr
    3.600,00 kr
    1 * 1 Each
       3.600,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    28-9035-29
    Opbevaring betingelser
    Enzyme mix to be stored at -70°C; all other components at -20°C.
    Thaw components on ice at 0 to 4°C during handling
  • 28-9035-31
    illustra™ TempliPhi™ Sequence Resolver Kit
    illustra™ TempliPhi™ Sequence Resolver Kit
    200
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 18.480,00 kr
    18.480,00 kr
    1 * 1 Each
       18.480,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    28-9035-31
    Opbevaring betingelser
    Enzyme mix to be stored at -70°C; all other components at -20°C.
    Thaw components on ice at 0 to 4°C during handling