Orders LinkContactUsLinkComponent
Test papers, lead acetate papers
Test papers, lead acetate papers
Test papers, lead acetate papers
Leverandør:  MACHEREY-NAGEL
Certifikater
Test papers, lead acetate papers
3 tilgængelige muligheder nedenfor
Product Details & Documents
These test papers allow quick and easy detection of hydrogen sulphide. This gas occurs in the processing of raw oil. It is toxic even in low concentrations. Therefore, critical points are carefully checked. Sulphide containing solutions also give a positive reaction.
  • Colour changes: White to brown-black
Lead acetate is toxic and the test paper needs to be declared as hazardous. The technically equivalent Sulphide test paper is a non-toxic and safe alternative.
Detection limit: Sulfide - 5 mg/L S²⁻
Color change: Sulfide - white → brown-black
Specifications for Product Family
Specifikationer
  • Catalog No:
  • MANA90744
  • MANA90745
  • MANA90746
  • Længde:
  • 75 mm
  • Bredde:
  • 7 mm
  • 7 mm
  • 10 mm
  • Shelf life:
  • 2 Years
  • 2 Years
  • 2 Years
Specifikationer
Product Family Options
Produktinformation
  • Område
    Tilgængelighed
    Price
  • MANA90744
    Bly(II)acetat test papir, 5 m
    Bly(II)acetat test papir, 5 m
    5 mg/l S²⁻
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 153,00 kr
    153,00 kr
    1 * 1 ROL
       153,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA90744
    Længde
    Bredde
    7 mm
    Shelf life
    2 Years
  • MANA90745
    Bly(II)acetat test papir, refil pakke 3×5 m
    Bly(II)acetat test papir, refil pakke 3×5 m
    5 mg/l S²⁻
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 3 ROL
    1 * 3 ROL - 169,00 kr
    169,00 kr
    1 * 3 ROL
       169,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA90745
    Længde
    Bredde
    7 mm
    Shelf life
    2 Years
  • MANA90746
    Bly(II)acetat test papir, strimler, 10 × 75mm
    Bly(II)acetat test papir, strimler, 10 × 75mm
    5 mg/l S²⁻
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 79,60 kr
    79,60 kr
    1 * 100 Items
       79,60 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MANA90746
    Længde
    75 mm
    Bredde
    10 mm
    Shelf life
    2 Years