Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, Trans-Anethole
 :   Merck
Supelco® Organic Reference Standard, Trans-Anethole
 :  Merck
Supelco® Organic Reference Standard, Trans-Anethole
Supelco® Organic Reference Standard, Trans-Anethole
  • GHS signal: GHS07, GHS09
 : P261 - P272 - P273 - P280 - P302 + P352 - P333 + P313
  • Catalog No:
  • 10368-1ML
  • 10368-5ML
  • Synonym(s):
  • 4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
  • 4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • Densitet:
  • 0,988 g/ml at 25 °C (lit.)
  • 0,988 g/ml at 25 °C (lit.)
  • CAS-nr:
  • 4180-23-8
  • 4180-23-8
  • MDL No.:
  • MFCD00009284
  • MFCD00009284
  • Formel:
  • CH₃CH=CHC₆H₄OCH₃
  • CH₃CH=CHC₆H₄OCH₃
  • Molekylvægt:
  • 148,20
  • 148,20
  • Melting point:
  • 20...21 °C (lit.)
  • 20...21 °C (lit.)
  • Kogepunkt:
  • 234...237 °C (lit.)
  • 234...237 °C (lit.)
  • Fareklassebeskrivelse:
  • H317 - H411
  • H317 - H411
  • Flammepunkt:
  • 101 °C
  • 101 °C
  • Shipping conditions:
  • STD
  • STD
  • Opbevaring betingelser:
  • 2...8 °C
  • 2...8 °C
Product Family Options
  • Beskrivelse
    Standard
    Pakning
    Pakn
  • trans-Anethole
    Analytical standard
    Glass bottle
    1 mL
    1 * 1 mL
    1 * 1 mL - 362,00 kr
    362,00 kr
    1 * 1 mL
       362,00 kr
    Specifications
    Cat. No.
    10368-1ML
    Synonym(s)
    4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
    M-Clarity quality level
    MQ100
    Densitet
    0,988 g/ml at 25 °C (lit.)
    CAS-nr
    4180-23-8
    MDL No.
    MFCD00009284
    Formel
    CH₃CH=CHC₆H₄OCH₃
    Molekylvægt
    148,20
    Melting point
    20...21 °C (lit.)
    Kogepunkt
    234...237 °C (lit.)
    Fareklassebeskrivelse
    H317 - H411
    Flammepunkt
    101 °C
    Shipping conditions
    STD
    Opbevaring betingelser
    2...8 °C
  • trans-Anethole
    Analytical standard
    Glass bottle
    5 mL
    1 * 5 mL
    1 * 5 mL - 1.410,00 kr
    1.410,00 kr
    1 * 5 mL
       1.410,00 kr
    Specifications
    Cat. No.
    10368-5ML
    Synonym(s)
    4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
    M-Clarity quality level
    MQ100
    Densitet
    0,988 g/ml at 25 °C (lit.)
    CAS-nr
    4180-23-8
    MDL No.
    MFCD00009284
    Formel
    CH₃CH=CHC₆H₄OCH₃
    Molekylvægt
    148,20
    Melting point
    20...21 °C (lit.)
    Kogepunkt
    234...237 °C (lit.)
    Fareklassebeskrivelse
    H317 - H411
    Flammepunkt
    101 °C
    Shipping conditions
    STD
    Opbevaring betingelser
    2...8 °C