Orbital molecular models
Orbital molecular models
Katalog # 761-0114
Marketsource
Leverandør: SPIRING ENTERPRISES
Gå til:
Product Details & Documents
Four models of molecular orbital configurations using benzene, ethane, ethene, and ethyne.
The models depict sigma and pi bonds and effects such as hybridisation and delocalised electrons.
Specifikationer
- Beskrivelse:Orbital molecular models