Orders LinkContactUsLinkComponent
Sample Containers, Sterile
Sample Containers, Sterile
Leverandør:   BURKLE
Sample Containers, Sterile
Leverandør:  BURKLE
Certifikater
Sample Containers, Sterile
Sample Containers, Sterile
3 tilgængelige muligheder nedenfor
Product Details & Documents
Sample box with tightly closing sealing cover.
  • Tightly closing sample container
  • With attached, vertically fixable snap-on lid
  • PP, transparent
The lid can be opened with one hand and its hinge angle is 90°. Liquid/condensate in the lid runs into an absorbing groove integrated in the lid, nothing is lost, and nothing can drip. The box is completely tight, has no crevices, and is therefore easy to fill and can be emptied without residues. It is autoclavable and suitable for deep freezing.
Product Family Options
Produktinformation
  • Kapacitet
    Udv.Ø
    Højde
    Steril
    Tilgængelighed
    Price
  • BURK6206-1100
    Graduated to 80 ml
    Graduated to 80 ml
    90 ml
    43 mm
    79 mm
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 350 Items
    1 * 350 Items - 2.280,00 kr
    2.280,00 kr
    1 * 350 Items
       2.280,00 kr
  • Graduated
    300 ml
    75 mm
    88 mm
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 240 Items
    1 * 240 Items - 4.240,00 kr
    4.240,00 kr
    1 * 240 Items
       4.240,00 kr
  • Graduated
    50 ml
    31 mm
    78 mm
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 650 Items
    1 * 650 Items
    Specifikationer:
    New Product