Glass Inserts for 96 Well Plates, Microsolv tech corp
Catalog # MICS95096-710
Supplier: MICROSOLV
Specifications
- Kapacitet:1,00 ml
- Beskrivelse:Glass inserts for 96 well plates
- Materiale:Borosilikatglas
- Pakn:250
Specifications
About this item
Glass inserts for 96 well plates, 1 ml, replacements. These borosilicate glass inserts are 1 ml and are replacements for 95096-1CP or 95096-3CP.
Ordering information: Plates and mats are sold separately.