Anti-PSA (Prostate Specific Antigen) Rabbit Monoclonal Antibody [clone: D2A8]
Katalog # CSTA5877S
Leverandør: Cell Signaling
Specifikationer
- Antistof type:Primary
- Antigen symbol:PSA (Prostate Specific Antigen)
- Clonality:Monoclonal
- Clone:D2A8
- Vært:Rabbit
- ImmunFluorescens:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sekvens:CSLLKNRFLRPGDDSS
- Opbevaringstemperatur:−20 °C
- Shipping temperature:Ambient
- Størrelse:100 µl
- Pakn:100 µl
Specifikationer
Om denne vare
Type: Primary Antibodies
Antigen: PSA (Prostate Specific Antigen)
Clonality: Monoclonal
Clone: D2A8
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human