Chemical Goggles
Chemical goggles, indirect vent
PRTWPS21CLR
: Portwest
PS21 are indirect ventilation goggles with acetate lens material, suitable for chemical use.
- Indirect ventilation
- Flexible PVC frame for ultra comfort
- Ultra-lightweight for long uses
- Non-magnetic construction; nickel- and ferrous-free
- Anti-fog and anti-scratch treatment coating
- Anti-scratch coating for added durability
- Retail bag aids presentation for retail sales
Superior anti-fog treatment (marked N on the lens) and ultra-soft frame ensure comfortable use for long working days.
- Beskrivelse:Chemical goggles, indirect vent