Cotton Bib Apron
Cotton bib apron, sort
Katalog # PRTWS840BKR
Leverandør: Portwest
Gå til:
Product Details & Documents
Made from 100% cotton fabric for added comfort and breathability cotton bib apron.
- Adjustable neck for a secure fit
- Anti-tangle ties for secure fastening
Cotton bib apron measure 720×950 mm.
Specifikationer
- Farve:sort
- Beskrivelse:Cotton bib apron