Gas Welding Goggles
Gas welding goggles, indirect ventilation, grøn
Katalog # PRTWPW60BGR
Leverandør: Portwest
Gå til:
Product Details & Documents
When using oxyacetylene for gas cutting or welding, the PW60 goggles offers high performance with a shade factor five.
- Indirect ventilation
- Flexible PVC frame for ultra comfort
- Stand-off position for intermittent use
- Adjustable elastic headband for a perfect fit
- Anti-scratch coating for added durability
- 99% UV protection
- Retail bag aids presentation for retail sales
A soft comfortable frame with flip up lens allows the goggle to be used for grinding operations. Indirect ventilation goggles are individually packaged.
Certifications: EN 169 (shade 5).
Specifikationer
- Farve:grøn
- Beskrivelse:Gas welding goggles, indirect ventilation