Pneumatic trough, pp, Nalgene®
Water
Katalog # 216-0428
Leverandør: Thermo Fisher Scientific
Pneumatic trough, pp, Nalgene®
Katalog # 216-0428
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Replace metal-type troughs with the polypropylene pneumatic trough.
- Lightweight and rustproof
- Gas tubing passes through a loop to prevent liquid from flowing into gas line
- Liquid inlet and outlet connections accept ¼" int.Ø × ³/₈" Ø (6,4×9,5 mm) tubing
This trough features tapered sides for nesting and a built-in gas inlet. There are no sharp corners or separate parts to lose. The built-in platform accommodates four 250 ml bottles.
Specifikationer
- Beskrivelse:Water
- Cat. No.:216-0428
- Miljøcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lav fremstillingspåvirkning